Anti-STK25 Rabbit Monoclonal Antibody [clone: EPR10306]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR10306] to STK25.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: STK25
Clonality: Monoclonal
Clone: EPR10306
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB157188-100U
- ABCA_AB157188-1ML
- Antigen Symbol:
- STK25
- STK25
- Antigen Name:
- SOK1
- SOK1
- Antigen Synonyms:
- YSK1|Serine/threonine-protein kinase 25|Ste20-like kinase|Sterile 20/oxidant stress-response kinase 1|SOK-1|Ste20/oxidant stress response kinase 1
- YSK1|Serine/threonine-protein kinase 25|Ste20-like kinase|Sterile 20/oxidant stress-response kinase 1|SOK-1|Ste20/oxidant stress response kinase 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR10306
- EPR10306
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 48112 Da
- 48112 Da
- Purification:
- Tissue culture supernatant
- Tissue culture supernatant
- Storage Temperature:
- Store at–20 ℃.
- Store at–20 ℃.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Specifications
Frequently Bought Together