Anti-AAV8+AAV3B Mouse Monoclonal Antibody [clone: HL2383]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [HL2383] to AAV8 + AAV3B.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: AAV8+AAV3B
Clonality: Monoclonal
Clone: HL2383
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Specifications
- Catalog No:
- ABCA_AB315827-100U
- ABCA_AB315827-20UG
- ABCA_AB315827-1MG
- Antigen Symbol:
- AAV8+AAV3B
- AAV8+AAV3B
- AAV8+AAV3B
- Antigen Name:
- Adeno-associated virus - 8
- Adeno-associated virus - 8
- Adeno-associated virus - 8
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- HL2383
- HL2383
- HL2383
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG3
- IgG3
- IgG3
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Dot, I-ELISA
- Dot, I-ELISA
- Dot, I-ELISA
Specifications
Frequently Bought Together