Anti-TRAP alpha/TRAPA Rabbit Polyclonal Antibody
ABCA_AB240562-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:TRAPA
- Antigen Symbol:TRAP alpha/TRAPA
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:PSEC0262|SSR1|Translocon-associated protein subunit alpha|TRAP-alpha|Signal sequence receptor subunit alpha|SSR-alpha
- Molecular Weight:32235 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human SSR1 aa 200 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Affinity purification
- Packaging:Vial
Rabbit polyclonal to TRAP alpha/TRAPA.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TRAP alpha/TRAPA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human