Anti-Thyrotropin Releasing Hormone (TRH) Rabbit Polyclonal Antibody
Catalog # ABCA_AB216601-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Pro-thyrotropin-releasing hormone
- Antigen Symbol:Thyrotropin Releasing Hormone (TRH)
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Pro-TRH|Prothyroliberin|TRH
- Molecular Weight:27404 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human TRH conjugated to Keyhole Limpet Haemocyanin. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Thyrotropin Releasing Hormone (TRH).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Thyrotropin Releasing Hormone (TRH)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human