Anti-RBM4 Rabbit Polyclonal Antibody
Catalog # ABCA_AB251923-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:RBM4A
- Antigen Symbol:RBM4
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:RNA-binding protein 4|Lark homolog|RNA-binding motif protein 4|RNA-binding motif protein 4a|hLark
- Molecular Weight:40314 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human RBM4 aa 300 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to RBM4.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RBM4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human