Anti-E2F3 Rabbit Polyclonal Antibody
ABCA_AB152126-100U
: Abcam
New Product
- Antibody Type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Transcription factor E2F3|E2F-3
- Molecular Weight:49162 Da
- Storage Temperature:+4 °C
- Concentration:0.28 mg/ml
- Shipping Temperature:Shipped at +4 °C. Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
- Immunogen:Recombinant Fragment Protein within Human E2F3 aa 150-450. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to E2F3.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: E2F3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human