Anti-Heme Oxygenase 1 Mouse Monoclonal Antibody [clone: HO-1-2] (FITC (Fluorescein Isothiocyanate))
ABCA_AB69545-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:HO
- Antigen Symbol:Heme Oxygenase 1
- Clonality:Monoclonal
- Clone:HO-1-2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:HO1|HMOX1|HO-1
- Storage Buffer:PBS
- Molecular Weight:32819 Da
- Storage Temperature:Store at 4 °C
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Native Full Length Protein corresponding to Rat Hmox1
- Tested Applications:Flow Cyt (Intra)
- Purification:Protein G affinity purification
- Packaging:Vial
FITC Mouse monoclonal [HO-1-2] to Heme Oxygenase 1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Heme Oxygenase 1
Clonality: Monoclonal
Clone: HO-1-2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human