Anti-NKR-P1C Rabbit Monoclonal Antibody [clone: EPR22990-31]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR22990-31] to NKR-P1C - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NKR-P1C
Clonality: Monoclonal
Clone: EPR22990-31
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- ABCA_AB289553-1MG
- ABCA_AB289553-100U
- Antigen Symbol:
- NKR-P1C
- NKR-P1C
- Antigen Name:
- CD161c
- CD161c
- Antigen Synonyms:
- Ly55c|Nkrp1c|Klrb1c|Killer cell lectin-like receptor subfamily B member 1C|CD161 antigen-like family member C|Lymphocyte antigen 55c|NK1.1|NKR-P1.9|Natural killer cell surface protein P1-40|Ly-55c|NKR-P1 40
- Ly55c|Nkrp1c|Klrb1c|Killer cell lectin-like receptor subfamily B member 1C|CD161 antigen-like family member C|Lymphocyte antigen 55c|NK1.1|NKR-P1.9|Natural killer cell surface protein P1-40|Ly-55c|NKR-P1 40
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR22990-31
- EPR22990-31
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 25077 Da
- 25077 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C.
- Store at +4 °C.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ICC/IF, IHC-Fr, IHC-P
- ICC/IF, IHC-Fr, IHC-P
Specifications
Frequently Bought Together