Order Entry
United States
ContactUsLinkComponent
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: SP185]
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: SP185]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-GCDFP 15 Rabbit Monoclonal Antibody [clone: SP185]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB183321-100U
  • ABCA_AB183321-1ML
  • ABCA_AB183321-500U
  • Antigen Symbol:
  • GCDFP 15
  • GCDFP 15
  • GCDFP 15
  • Antigen Name:
  • GCDFP15
  • GCDFP15
  • GCDFP15
  • Antigen Synonyms:
  • GPIP4|PIP|Prolactin-inducible protein|Gross cystic disease fluid protein 15|Prolactin-induced protein|Secretory actin-binding protein|gp17|GCDFP-15|SABP
  • GPIP4|PIP|Prolactin-inducible protein|Gross cystic disease fluid protein 15|Prolactin-induced protein|Secretory actin-binding protein|gp17|GCDFP-15|SABP
  • GPIP4|PIP|Prolactin-inducible protein|Gross cystic disease fluid protein 15|Prolactin-induced protein|Secretory actin-binding protein|gp17|GCDFP-15|SABP
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • SP185
  • SP185
  • SP185
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular Weight:
  • 16572 Da
  • 16572 Da
  • 16572 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • 4 °C
  • Tested Applications:
  • IHC-P
  • IHC-P
  • IHC-P

Specifications

Frequently Bought Together