Anti-GluK5 Rabbit Polyclonal Antibody
Catalog # ABCA_AB67408-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:GRIK2
- Antigen Symbol:GluK5
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:GRIK5|Excitatory amino acid receptor 2|Glutamate receptor KA-2|EAA2|KA2
- Storage Buffer:PBS
- Molecular Weight:109265 Da
- Storage Temperature:Store at 4 °C
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human GRIK5. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to GluK5.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GluK5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human