Anti-MDM2 Rabbit Monoclonal Antibody [clone: EPR22256-98]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR22256-98] to MDM2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: EPR22256-98
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Specifications
- Catalog No:
- ABCA_AB259265-100U
- ABCA_AB259265-1ML
- ABCA_AB259265-20UL
- Antigen Symbol:
- MDM2
- MDM2
- MDM2
- Antigen Name:
- E3 ubiquitin-protein ligase Mdm2
- E3 ubiquitin-protein ligase Mdm2
- E3 ubiquitin-protein ligase Mdm2
- Antigen Synonyms:
- Double minute 2 protein|Oncoprotein Mdm2|RING-type E3 ubiquitin transferase Mdm2|p53-binding protein Mdm2|Hdm2
- Double minute 2 protein|Oncoprotein Mdm2|RING-type E3 ubiquitin transferase Mdm2|p53-binding protein Mdm2|Hdm2
- Double minute 2 protein|Oncoprotein Mdm2|RING-type E3 ubiquitin transferase Mdm2|p53-binding protein Mdm2|Hdm2
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- EPR22256-98
- EPR22256-98
- EPR22256-98
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 55233 Da
- 55233 Da
- 55233 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- ICC/IF
- ICC/IF
- ICC/IF
Specifications
Frequently Bought Together