Order Entry
United States
ContactUsLinkComponent
Anti-Uroplakin III Rabbit Monoclonal Antibody [clone: SP73]
Anti-Uroplakin III Rabbit Monoclonal Antibody [clone: SP73]
Supplier:  Abcam
New Product
Certificates
Anti-Uroplakin III Rabbit Monoclonal Antibody [clone: SP73]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB93721-500UL
  • ABCA_AB93721-100UL
  • ABCA_AB93721-1ML
  • Antigen Symbol:
  • Uroplakin III
  • Uroplakin III
  • Uroplakin III
  • Antigen Name:
  • UPK3
  • UPK3
  • UPK3
  • Antigen Synonyms:
  • UPK3A|Uroplakin-3a|UP3a|UPIII
  • UPK3A|Uroplakin-3a|UP3a|UPIII
  • UPK3A|Uroplakin-3a|UP3a|UPIII
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • SP73
  • SP73
  • SP73
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Epitope:
  • C-terminus.
  • C-terminus.
  • C-terminus.
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular Weight:
  • 30670 Da
  • 30670 Da
  • 30670 Da
  • Purification:
  • Protein A/G Affinity purification
  • Protein A/G Affinity purification
  • Protein A/G Affinity purification
  • Storage Buffer:
  • PBS, BSA
  • PBS, BSA
  • PBS, BSA
  • Storage Temperature:
  • Shipped at +4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Shipped at +4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Shipped at +4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Shipping Temperature:
  • +4 °C
  • +4 °C
  • +4 °C
  • Tested Applications:
  • IHC-P
  • IHC-P
  • IHC-P

Specifications

Frequently Bought Together