Order Entry
United States
ContactUsLinkComponent
Anti-IRS1 Rabbit Monoclonal Antibody [clone: EPR23651-4]
Anti-IRS1 Rabbit Monoclonal Antibody [clone: EPR23651-4]
Catalog # ABCA_AB280019-100U
Supplier:  Abcam
New Product
Anti-IRS1 Rabbit Monoclonal Antibody [clone: EPR23651-4]
Catalog # ABCA_AB280019-100U
Supplier:  Abcam
Supplier Number:  AB280019-100UG
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Insulin receptor substrate 1
  • Antigen Symbol:
    IRS1
  • Clonality:
    Monoclonal
  • Clone:
    EPR23651-4
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Size:
    100 µg
  • Form:
    Liquid
  • Antigen Synonyms:
    IRS-1
  • Molecular Weight:
    131591 Da
  • Storage Temperature:
    Store at +4 °C.
  • Shipping Temperature:
    Shipped at +4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information.
  • Tested Applications:
    sELISA
  • Purification:
    Protein A Affinity purification
  • Packaging:
    Vial

Specifications

About this item

Rabbit monoclonal [EPR23651-4] to IRS1 - BSA and Azide free (Capture).

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: EPR23651-4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human