Anti-PP2A alpha + beta Rabbit Monoclonal Antibody [clone: Y119] (Alexa Fluor® 488)
ABCA_AB203957-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
- Antigen Symbol:PP2A alpha + beta
- Clonality:Monoclonal
- Clone:Y119
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Epitope:ab203957 reacts with an epitope located in the C terminal region of PP2A alpha and beta
- Form:Liquid
- Antigen Synonyms:PP2A-alpha|Replication protein C|RP-C|PPP2CA
- Molecular Weight:35594 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Stable for 12 months at –20 °C. Store In the Dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Protein A Affinity purification
- Packaging:Vial
Alexa Fluor® 488 Rabbit monoclonal [Y119] to PP2A alpha + beta.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PP2A alpha + beta
Clonality: Monoclonal
Clone: Y119
Conjugation: Alexa Fluor® 488
Epitope: ab203957 reacts with an epitope located in the C terminal region of PP2A alpha and beta
Host: Rabbit
Isotype: IgG
Reactivity: Human