Anti-Reptin/TIP49B/RUVB2 Rabbit Monoclonal Antibody [clone: EPR4146]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR4146] to Reptin/TIP49B/RUVB2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Reptin/TIP49B/RUVB2
Clonality: Monoclonal
Clone: EPR4146
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB108962-1ML
- ABCA_AB108962-100U
- Antigen Symbol:
- Reptin/TIP49B/RUVB2
- Reptin/TIP49B/RUVB2
- Antigen Name:
- INO80J
- INO80J
- Antigen Synonyms:
- TIP48|TIP49B|CGI-46|RUVBL2|RuvB-like 2|48 kDa TATA box-binding protein-interacting protein|51 kDa erythrocyte cytosolic protein|INO80 complex subunit J|Repressing pontin 52|TIP60-associated protein 54-beta|48 kDa TBP-interacting protein|ECP-51|Reptin 52|TAP54-beta
- TIP48|TIP49B|CGI-46|RUVBL2|RuvB-like 2|48 kDa TATA box-binding protein-interacting protein|51 kDa erythrocyte cytosolic protein|INO80 complex subunit J|Repressing pontin 52|TIP60-associated protein 54-beta|48 kDa TBP-interacting protein|ECP-51|Reptin 52|TAP54-beta
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR4146
- EPR4146
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 51157 Da
- 51157 Da
- Purification:
- Tissue culture supernatant
- Tissue culture supernatant
- Storage Buffer:
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- Storage Temperature:
- Store at −20 °C. Stable for 12 months at −20 °C.
- Store at −20 °C. Stable for 12 months at −20 °C.
- Shipping Temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together