Order Entry
United States
ContactUsLinkComponent
Anti-GPCR GPR48/LGR4 Mouse Monoclonal Antibody [clone: 6G8.B3.G5.C3]
Anti-GPCR GPR48/LGR4 Mouse Monoclonal Antibody [clone: 6G8.B3.G5.C3]
  ABCA_AB106089-100U
 :  Abcam
New Product
Anti-GPCR GPR48/LGR4 Mouse Monoclonal Antibody [clone: 6G8.B3.G5.C3]
  ABCA_AB106089-100U
 :  Abcam
 :  AB106089-100UG
New Product

 

Mouse monoclonal [6G8.B3.G5.C3] to GPCR GPR48/LGR4.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GPCR GPR48/LGR4
Clonality: Monoclonal
Clone: 6G8.B3.G5.C3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human