Order Entry
United States
ContactUsLinkComponent
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EPR5371]
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EPR5371]
Supplier:  Abcam
New Product
Certificates
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EPR5371]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB124808-100U
  • ABCA_AB124808-1ML
  • Antigen Symbol:
  • Antithrombin III/ATIII
  • Antithrombin III/ATIII
  • Antigen Name:
  • AT3
  • AT3
  • Antigen Synonyms:
  • PRO0309|SERPINC1|Antithrombin-III|ATIII|Serpin C1
  • PRO0309|SERPINC1|Antithrombin-III|ATIII|Serpin C1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR5371
  • EPR5371
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 52602 Da
  • 52602 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Storage Temperature:
  • Store at –20 °C. Stable for 12 months at –20 °C.
  • Store at –20 °C. Stable for 12 months at –20 °C.
  • Shipping Temperature:
  • Shipped at +4 °C
  • Shipped at +4 °C

Specifications

Frequently Bought Together