Anti-BAZ2B Rabbit Monoclonal Antibody [clone: EPR20909-88]
: Abcam
New Product
Rabbit monoclonal [EPR20909-88] to BAZ2B.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: BAZ2B
Clonality: Monoclonal
Clone: EPR20909-88
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- ABCA_AB234420-100U
- ABCA_AB234420-1ML
- Antigen Symbol:
- BAZ2B
- BAZ2B
- Antigen Name:
- Bromodomain adjacent to zinc finger domain protein 2B
- Bromodomain adjacent to zinc finger domain protein 2B
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR20909-88
- EPR20909-88
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
Frequently Bought Together