Anti-LRRK2 (phospho S935) Rabbit Monoclonal Antibody [clone: UDD2 10(12)]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [UDD2 10(12)] to LRRK2 (phospho S935) - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: LRRK2
Clonality: Monoclonal
Clone: UDD2 10(12)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- ABCA_AB172382-100U
- ABCA_AB172382-1MG
- Antigen Symbol:
- LRRK2
- LRRK2
- Antigen Name:
- PARK8
- PARK8
- Antigen Synonyms:
- Leucine-rich repeat serine/threonine-protein kinase 2|Dardarin
- Leucine-rich repeat serine/threonine-protein kinase 2|Dardarin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- UDD2 10(12)
- UDD2 10(12)
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Modification:
- phospho S935
- phospho S935
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 286103 Da
- 286103 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together