Order Entry
United States
ContactUsLinkComponent
Anti-SLCO1B3/OATP1B3 Mouse Monoclonal Antibody [clone: CL3770]
Anti-SLCO1B3/OATP1B3 Mouse Monoclonal Antibody [clone: CL3770]
Catalog # ABCA_AB242368-100U
Supplier:  Abcam
New Product
undefined
Anti-SLCO1B3/OATP1B3 Mouse Monoclonal Antibody [clone: CL3770]
Catalog # ABCA_AB242368-100U
Supplier:  Abcam
Supplier Number:  AB242368-100UL
New Product

Specifications

  • Size:
    100 µl
  • Packaging:
    Vial

Specifications

About this item

Mouse monoclonal [CL3770] to SLCO1B3/OATP1B3.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SLCO1B3/OATP1B3
Clonality: Monoclonal
Clone: CL3770
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human