Anti-RSK1 p90 Rabbit Monoclonal Antibody [clone: E4]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [E4] to RSK1 p90 - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RSK1 p90
Clonality: Monoclonal
Clone: E4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Mouse
Specifications
- Catalog No:
- ABCA_AB239805-1MG
- ABCA_AB239805-100U
- Antigen Symbol:
- RSK1 p90
- RSK1 p90
- Antigen Name:
- MAPKAPK1A
- MAPKAPK1A
- Antigen Synonyms:
- RSK1|RPS6KA1|Ribosomal protein S6 kinase alpha-1|S6K-alpha-1|90 kDa ribosomal protein S6 kinase 1|MAP kinase-activated protein kinase 1a|Ribosomal S6 kinase 1|p90-RSK 1|p90RSK1|p90S6K|MAPK-activated protein kinase 1a|MAPKAP kinase 1a|MAPKAPK-1a|RSK-1
- RSK1|RPS6KA1|Ribosomal protein S6 kinase alpha-1|S6K-alpha-1|90 kDa ribosomal protein S6 kinase 1|MAP kinase-activated protein kinase 1a|Ribosomal S6 kinase 1|p90-RSK 1|p90RSK1|p90S6K|MAPK-activated protein kinase 1a|MAPKAP kinase 1a|MAPKAPK-1a|RSK-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- E4
- E4
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 82723 Da
- 82723 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Temperature:
- Store at +4 °C. Do not freeze.
- Store at +4 °C. Do not freeze.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together