Anti-Cdk6 (phospho Y13) Rabbit Polyclonal Antibody
Catalog # ABCA_AB131439-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CDKN6
- Antigen Symbol:Cdk6
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Cyclin-dependent kinase 6|Cell division protein kinase 6|Serine/threonine-protein kinase PLSTIRE
- Modification:phospho Y13
- Molecular Weight:36938 Da
- Storage Temperature:Store at –20 °C. Stable for 12 months at –20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human CDK6 phospho Y13 conjugated to Keyhole Limpet Haemocyanin. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Cdk6 (phospho Y13).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cdk6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human