Anti-sPLA2-X Rabbit Monoclonal Antibody [clone: EPR11202]
: Abcam
New Product
Rabbit monoclonal [EPR11202] to sPLA2-X.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: sPLA2-X
Clonality: Monoclonal
Clone: EPR11202
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB166634-1ML
- ABCA_AB166634-100U
- Antigen Symbol:
- sPLA2-X
- sPLA2-X
- Antigen Name:
- Group 10 secretory phospholipase A2
- Group 10 secretory phospholipase A2
- Antigen Synonyms:
- Group X secretory phospholipase A2|Phosphatidylcholine 2-acylhydrolase 10|GX sPLA2|PLA2G10
- Group X secretory phospholipase A2|Phosphatidylcholine 2-acylhydrolase 10|GX sPLA2|PLA2G10
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR11202
- EPR11202
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 18153 Da
- 18153 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Shipped at +4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipped at +4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- Flow Cyt (Intra), IHC-P
- Flow Cyt (Intra), IHC-P
Frequently Bought Together