Anti-Clathrin heavy chain Mouse Monoclonal Antibody [clone: X22]
ABCA_AB2731-100UL
: Abcam
New Product
- Size:100 µl
- Packaging:Vial
Mouse monoclonal [X22] to Clathrin heavy chain.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Clathrin heavy chain
Clonality: Monoclonal
Clone: X22
Conjugation:
Epitope: Electron microscopy and proteolysis mapping demonstrate that binding occurs towards the central hub of the triskelion, N-terminal to the light chain binding regions.
Host: Mouse
Isotype: IgG1
Reactivity: Human;Xenopus laevis