Anti-Rho guanine exchange factor 16/NBR Rabbit Monoclonal Antibody [clone: EPR9950]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR9950] to Rho guanine exchange factor 16/NBR.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Rho guanine exchange factor 16/NBR
Clonality: Monoclonal
Clone: EPR9950
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Specifications
- Catalog No:
- ABCA_AB154785-100U
- ABCA_AB154785-1ML
- Antigen Symbol:
- Rho guanine exchange factor 16/NBR
- Rho guanine exchange factor 16/NBR
- Antigen Name:
- EPHEXIN4
- EPHEXIN4
- Antigen Synonyms:
- NBR|ARHGEF16|Rho guanine nucleotide exchange factor 16|Ephexin-4
- NBR|ARHGEF16|Rho guanine nucleotide exchange factor 16|Ephexin-4
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR9950
- EPR9950
- Reactivity:
- Mouse, Human
- Mouse, Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 80105 Da
- 80105 Da
- Purification:
- Tissue culture supernatant
- Tissue culture supernatant
- Storage Temperature:
- Store at -20ºC
- Store at -20ºC
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together