Order Entry
United States
ContactUsLinkComponent
Anti-0 Mouse 0 Antibody [clone: 7E10G10] (Alexa Fluor® 594)
Anti-0 Mouse 0 Antibody [clone: 7E10G10] (Alexa Fluor® 594)
Catalog # ABCA_AB178002-100U
Supplier:  Abcam
New Product
undefined
Anti-0 Mouse 0 Antibody [clone: 7E10G10] (Alexa Fluor® 594)
Catalog # ABCA_AB178002-100U
Supplier:  Abcam
Supplier Number:  AB178002-100UL
New Product

Specifications

  • Size:
    100 µl
  • Packaging:
    Vial

Specifications

About this item

Alexa Fluor® 594 Mouse IgG2b, kappa monoclonal [7E10G10] - Isotype Control.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: IgG2b
Clonality: Monoclonal
Clone: 7E10G10
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: