Anti-Histone H2B Mouse Monoclonal Antibody [clone: mAbcam 52484]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [mAbcam 52484] to Histone H2B - ChIP Grade.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H2B
Clonality: Monoclonal
Clone: mAbcam 52484
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse; Rat; Human
Specifications
- Catalog No:
- ABCA_AB52484-100UG
- ABCA_AB52484-1MG
- Antigen Symbol:
- Histone H2B
- Histone H2B
- Antigen Name:
- H2BFF
- H2BFF
- Antigen Synonyms:
- HIST1H2BB|H2BC3|Histone H2B type 1-B|H2B-clustered histone 3|Histone H2B.1|Histone H2B.f|H2B/f
- HIST1H2BB|H2BC3|Histone H2B type 1-B|H2B-clustered histone 3|Histone H2B.1|Histone H2B.f|H2B/f
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- mAbcam 52484
- mAbcam 52484
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 13950 Da
- 13950 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Buffer:
- PBS
- PBS
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together