Anti-PHD1/prolyl hydroxylase Rabbit Monoclonal Antibody [clone: EPR2745]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR2745] to PHD1/prolyl hydroxylase.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PHD1/prolyl hydroxylase
Clonality: Monoclonal
Clone: EPR2745
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB108980-100U
- ABCA_AB108980-1ML
- Antigen Symbol:
- PHD1/prolyl hydroxylase
- PHD1/prolyl hydroxylase
- Antigen Name:
- EIT6
- EIT6
- Antigen Synonyms:
- EGLN2|Prolyl hydroxylase EGLN2|Egl nine homolog 2|Estrogen-induced tag 6|HPH-3|Hypoxia-inducible factor prolyl hydroxylase 1|Prolyl hydroxylase domain-containing protein 1|EIT-6|HIF-PH1|HIF-prolyl hydroxylase 1|HPH-1|PHD1
- EGLN2|Prolyl hydroxylase EGLN2|Egl nine homolog 2|Estrogen-induced tag 6|HPH-3|Hypoxia-inducible factor prolyl hydroxylase 1|Prolyl hydroxylase domain-containing protein 1|EIT-6|HIF-PH1|HIF-prolyl hydroxylase 1|HPH-1|PHD1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR2745
- EPR2745
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 43650 Da
- 43650 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Buffer:
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together