Anti-ATP5C1 Mouse Monoclonal Antibody [clone: 2A1AA11]
Catalog # ABCA_AB119686-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:ATP5C
- Antigen Symbol:ATP5C1
- Clonality:Monoclonal
- Clone:2A1AA11
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:ATP5CL1|ATP5F1C|ATP synthase F1 subunit gamma|F-ATPase gamma subunit
- Molecular Weight:32996 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Tested Applications:ICC/IF, IHC-P
- Purification:Ammonium Sulphate Precipitation
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [2A1AA11] to ATP5C1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATP5C1
Clonality: Monoclonal
Clone: 2A1AA11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human; Mouse; Rat; Bovine