Anti-IgG Donkey Polyclonal Antibody (Alexa Fluor® 680)
Catalog # ABCA_AB186698-500U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Secondary
- Antigen Name:Igh-4
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Donkey
- Western Blot:Yes
- Size:500 µg
- Form:Liquid
- Antigen Synonyms:Ighg1|Ig gamma-1 chain C region secreted form
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Stable for 12 months at −20 °C. Store in the dark.
- Concentration:2 mg/ml
- Shipping Temperature:4 °C
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Donkey Anti-Mouse IgG H&L (Alexa Fluor® 680) preadsorbed.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 680
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse