Anti-MDM2 Mouse Monoclonal Antibody [clone: 2A10]
Catalog # ABCA_AB16895-50UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:E3 ubiquitin-protein ligase Mdm2
- Antigen Symbol:MDM2
- Clonality:Monoclonal
- Clone:2A10
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Epitope:Within amino acids 294 - 339
- Form:Liquid
- Antigen Synonyms:Double minute 2 protein|Oncoprotein Mdm2|RING-type E3 ubiquitin transferase Mdm2|p53-binding protein Mdm2|Hdm2
- Storage Buffer:Sodium phosphate, gelatin
- Molecular Weight:55233 Da
- Storage Temperature:Store at 4 °C (stable for up to 12 months). Do not freeze.
- Concentration:0.1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Full Length Protein corresponding to Human MDM2 aa 250 - 350. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC, ICC/IF, IHC-P
- Purification:Tissue culture supernatant
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [2A10] to MDM2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: 2A10
Conjugation:
Epitope: Within amino acids 294 - 339
Host: Mouse
Isotype: IgG2a
Reactivity: Human