Order Entry
United States
ContactUsLinkComponent
Anti-SC35 Mouse Monoclonal Antibody [clone: SC-35]
Anti-SC35 Mouse Monoclonal Antibody [clone: SC-35]
Catalog # ABCA_AB264431-100U
Supplier:  Abcam
New Product
Anti-SC35 Mouse Monoclonal Antibody [clone: SC-35]
Catalog # ABCA_AB264431-100U
Supplier:  Abcam
Supplier Number:  AB264431-100UG
New Product

About this item

Mouse monoclonal [SC-35] to SC35 - BSA and Azide free.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GOLPH3/MIDAS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse