Anti-FLCN Rabbit Monoclonal Antibody [clone: EPNCIR147]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPNCIR147] to FLCN.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FLCN
Clonality: Monoclonal
Clone: EPNCIR147
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB124885-100U
- ABCA_AB124885-1ML
- Antigen Symbol:
- FLCN
- FLCN
- Antigen Name:
- BHD
- BHD
- Antigen Synonyms:
- Folliculin|BHD skin lesion fibrofolliculoma protein|Birt-Hogg-Dube syndrome protein
- Folliculin|BHD skin lesion fibrofolliculoma protein|Birt-Hogg-Dube syndrome protein
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPNCIR147
- EPNCIR147
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 64473 Da
- 64473 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at –20 °C. Stable for 12 months at –20 °C.
- Store at –20 °C. Stable for 12 months at –20 °C.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
Specifications
Frequently Bought Together