Anti-CHREBP Rabbit Polyclonal Antibody
Catalog # ABCA_AB92809-100UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:BHLHD14
- Antigen Symbol:CHREBP
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:MIO|WBSCR14|MLXIPL|Carbohydrate-responsive element-binding protein|Class D basic helix-loop-helix protein 14|MLX interactor|MLX-interacting protein-like|WS basic-helix-loop-helix leucine zipper protein|Williams-Beuren syndrome chromosomal region 14 protein|WS-bHLH
- Storage Buffer:Tris citrate/phosphate
- Molecular Weight:93073 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human MLXIPL aa 800 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to CHREBP.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CHREBP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human