Anti-Laminin alpha 5/LAMA5 Rabbit Monoclonal Antibody [clone: EPR18919]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR18919] to Laminin alpha 5/LAMA5.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Laminin alpha 5/LAMA5
Clonality: Monoclonal
Clone: EPR18919
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB184330-100U
- ABCA_AB184330-1ML
- Antigen Symbol:
- Laminin alpha 5/LAMA5
- Laminin alpha 5/LAMA5
- Antigen Name:
- KIAA0533
- KIAA0533
- Antigen Synonyms:
- KIAA1907|LAMA5|Laminin subunit alpha-5|Laminin-10 subunit alpha|Laminin-11 subunit alpha|Laminin-15 subunit alpha
- KIAA1907|LAMA5|Laminin subunit alpha-5|Laminin-10 subunit alpha|Laminin-11 subunit alpha|Laminin-15 subunit alpha
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR18919
- EPR18919
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 399737 Da
- 399737 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together