Dr. Ehrenstorfer 4-n-Octylphenol 13C6 100 µg/ml in Isooctane
Catalog # 77976-038
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Isooctane
- Description:4-n-Octylphenol 13C6 100 µg/ml in Isooctane
- Size:1 ml
- Shipping Conditions:20 °C
- Formula:¹³C₆C₈H₂₂O
- Formula Weight:212,28 g/mol
- Storage Temperature:20 °C
- Certification:ISO 17034