Orders LinkContactUsLinkComponent
Phosphate Comparator Flat
Phosphate Comparator Flat
Stannous Chloride
Catalog # 77674-676
Supplier:   CHEMetrics
Supplier #:  C-8510
Phosphate Comparator Flat
Catalog # 77674-676
Supplier:  CHEMetrics
CAS Number:  
Phosphate Comparator Flat
Phosphate Comparator Flat
Stannous Chloride
CAS Number:  
Price
$29.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The C-8510 Flat Phosphate Comparator is a component of K-8510 and K-8520D poshpate test kits. It is designed to be compared against the color formed in R-8510 Phosphate, ortho CHEMets® Refill. The comparatorandapos;s range is applicable for many industrial water, wastewater, and environmental water quality applications. The color standards contain blue dye mixtures that are formulated to match the colors produced by the test ampoules. Range: 1 to 10.0, 10 to 100 ppm.
Documents
Specifications
Specifications
  • Chemical method:
    Stannous Chloride