Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Density 15 °C (Nominal: 1.0248 g/ml @ 15 °C)
Paragon Scientific Density 15 °C (Nominal: 1.0248 g/ml @ 15 °C)
Catalog # 77975-654
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Density 15 °C (Nominal: 1.0248 g/ml @ 15 °C)
Catalog # 77975-654
Supplier:  LGC Standards
Supplier Number:  ALK-DEN15-10-500ML
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Density 15 °C (Nominal: 1.0248 g/ml @ 15 °C), Standard type: Multi-component
  • Size:
    500 ml

Specifications

About this item

  • Shipping conditions: Room temperature