Paragon Scientific Vapour Pressure ASTM D5191: DVPE at 46.9 kPa, Ptot at 52.5 kPa
Supplier: LGC Standards
New Product
Certificates
About this item
- Shipping conditions: Room temperature
Specifications
- Storage Temperature:RT
Specifications
Frequently Bought Together