Dr. Ehrenstorfer PFAS-C-ES Mixture 2 µg/ml in Methanol:Water
77976-056
New Product
:
- Matrix:Methanol:Water 96:4
- Description:PFAS-C-ES Mixture 2 µg/ml in Methanol:Water
- Size:1.2 ml
- Shipping Conditions:20 °C
- Storage Temperature:−18 °C
- Certification:ISO 17034