TRC Perfluorohexanesulfonic Acid (Technical Grade)
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:355-46-4
Specifications
Frequently Bought Together
J.T.Baker®, Syringe Filters
Supplier: Avantor
Acrodisc® Syringe Filters, 25 mm
Supplier: Cytiva
VWR® Syringe Filters
Supplier: VWR International
EZBio® Vent Filter, 0.22 µm, Nonsterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
Diamond® PureFlow™ PTFE Syringe Filters, Non Sterile, Globe Scientific
Supplier: Globe Scientific
Minisart® Acticosart Syringe Filters, Sartorius
Supplier: Sartorius
Acrodisc® Syringe Filters for Prefiltration, Cytiva
Supplier: Cytiva
Target2™ Nylon Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
Titan3™ PVDF Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
FILTER SYS 150ML PES 0.10UM 50MM S CS12
Supplier: VistaLab
Minisart® RC Syringe Filters
Supplier: Sartorius
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
BOT TP FILT PP 1000ML NYLN .45UM NS CS12
Supplier: VistaLab
Supor® Membrane Disc Filters
Supplier: Cytiva