TRC rac-Clopidogrel Hydrochloride
New Product
- CAS No.:144750-52-7
Frequently Bought Together
Ward's® Prepared Sabouraud Dextrose Media
: Culture Media & Supplies
MSA Mannitol Salt Agar Plates
: Culture Media & Supplies
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
Manual Crimpers and Decappers, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
: Upchurch Scientific
Titration Cup, Dyn-A-Med
: Dyn-A-Med
Detectors for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
J.T.Baker®, Phase Separation Paper
: AVANTOR PERFORMANCE MATERIAL LLC
pH Gradient Buffers
: Thermo Scientific
CrossLab Instrument Supplies for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Captiva Syringe Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Bond Elut™ Adapters and Caps, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Skimmer Cones for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Heat Exchanger for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)