Paragon Scientific Type N100HT, High Temperature Viscosity (Ranges: 100 to 150 °C)
Catalog # 77975-897
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Type N100HT, high temperature viscosity (Ranges: 100 - 150 °C)
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component