Paragon Scientific Density 100 °C (Nominal: 0.7645 g/ml @ 100 °C)
Supplier: LGC Standards
New Product
Certificates
Paragon Scientific Density 100 °C (Nominal: 0.7645 g/ml @ 100 °C)
Supplier: LGC Standards
New Product
About this item
- Shipping conditions: Room temperature
Frequently Bought Together
AGILENT 7850 7900 8900 BRACKET ASSEMBLY
Supplier: Glass Expansion Inc
CrossLab Sample Trays for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX Ion Exchange, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
Sulfur Chemiluminescence Detector Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
Guard Cartridge Holder, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
In-line Gas Purifiers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Nebulizer Supplies for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab GC Fittings, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)