Order Entry
United States
ContactUsLinkComponent
Paragon Scientific Relative Density 20 °C (Nominal: 0.8452 @ 20 °C)
Paragon Scientific Relative Density 20 °C (Nominal: 0.8452 @ 20 °C)
  77975-954
New Product
 :  
undefined
Paragon Scientific Relative Density 20 °C (Nominal: 0.8452 @ 20 °C)
  77975-954
 :  ALK-RDEN20-05
New Product
 :  

 

  • Pack type:
    Glass bottle
  • Description:
    Relative density 20 °C (Nominal: 0.8452 @ 20 °C), Standard type: Single component
  • Size:
    500 ml

 

 

  • Shipping conditions: Room temperature