Paragon Scientific Cone and Plate (Nominal: 500 mPas @ 25 °C), Viscosity
Catalog # 77975-468
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Cone and plate (nominal: 500 mPas @ 25 °C), viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component