Dr. Ehrenstorfer Microplastics Polypropylene PP nominal 20 μm) (Dv50 = 40 μm)
Catalog # 77976-079
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:9003-07-0
- Description:Microplastics polypropylene PP nominal 20 μm (Dv50 = 40 μm; Dn50 = 6 μm)
- Size:50 mg
- Shipping Conditions:20 °C
- Formula:n(C₃H₆)
- Formula Weight:42,08 g/mol
- Storage Temperature:20 °C
- Certification:ISO/IEC 17025