Paragon Scientific Density 15 °C (Nominal: 1.1962 g/ml @ 15 °C)
Supplier: LGC Standards
New Product
Certificates
About this item
- Shipping conditions: Room temperature
Frequently Bought Together
CrossLab Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Autosampler Valve Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sample Trays for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Vacuum Pump Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Burners and Supplies for Agilent Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Cleaning and Maintenance Supplies for LC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Stay Safe Caps & Accessories for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab GC Fittings, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
CrossLab Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)