Order Entry
United States
ContactUsLinkComponent
Nitrite Instrumental Test 0 to 1 ppm
Nitrite Instrumental Test 0 to 1 ppm
Catalog # 77674-832
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Nitrite Instrumental Test 0 to 1 ppm
Catalog # 77674-832
Supplier:  CHEMetrics
Supplier Number:  K-7003
New Product
CAS Number:  

Specifications

  • Chemical method:
    Azo dye formation
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water

Specifications

About this item

  • Range: 0 to 1.00 ppm as N Method: Azo dye formation
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, 25 ml sample cup, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.