Paragon Scientific Density 25 °C (Nominal: 1.0222 g/ml @ 25 °C)
Catalog # 77975-692
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Density 25 °C (Nominal: 1.0222 g/ml @ 25 °C), Standard type: Multi-component
- Size:500 ml
Specifications
About this item
- Shipping conditions: Room temperature